9240-11-011 - Rev 0 - Hotplate Stirrer, HPS RT2 Basic - Operation Manual
Description:
Operation manual
Environment:
Hotplate Stirrer, HPS RT2 Basic:
Item/Model | HP RT2 Basic |
Dimension (W x D x H, mm) | 161 x 290 x 100 |
Top plate dimension (Ø) | 140 mm (5.5”) |
Maximum heating capacity | 600W |
Current consumption (230V) | 3A |
Current consumption (120V) | 5A |
Attachment(s):
File | Last Modified |
---|---|
9240-11-011 - Rev 0 - Hotplate Stirrer, HPS RT2 Basic - Operation Manual.pdf | December 29, 2021 |