096-3000-00 - Rev D - ValuPump Vacuum Pump - Instruction Manual
The instruction Manual provides detailed installation, operation, and maintenance guidelines for the Thermo ValuPump models VLP80, VLP120, VLP200, and VLP285 Rotary Vane Pumps. It covers technical specifications, start-up procedures, achieving ultimate vacuum, handling condensable vapors, and comprehensive maintenance tasks including oil replacement and cleaning. Essential safety instructions are highlighted to ensure proper use and maintenance.
Description:
This manual provides installation, operation and maintenance instructions for the Thermo ValuPump VLP80, VLP120, VLP200 and VLP285 Rotary Vane Pumps.
Environment:
ValuPump Vacuum Pump, models:
| VLP80 | VLP120 | VLP200 | VLP285 | |
| Motor Power: | ||||
| 120V, 60 Hz (Watts) | 300 | 300 | 550 | 550 |
| Start-up Current (Amps) | 31.5 | 31.5 | 34.0 | 34.0 |
| Fuse Rating, (Amps) | 20 | 20 | 25 | 25 |
| 230V, 50 Hz (Watts) | 250 | 250 | 450 | 450 |
| Start-Up Current (Amps) | 15.6 | 15.6 | 18.0 | 18.0 |
| Fuse Rating, (Amps) | 10 | 10 | 13 | 13 |
Attachment(s):
| File | Last Modified |
|---|---|
| 096-3000-00 - Rev D - ValuPump Vacuum Pump - Instruction Manual.pdf | January 03, 2022 |