104-3001-00 - Rev E - Savant DNA110 SpeedVac Concentrator - Instruction Manual
The instruction manual provides comprehensive guidance on the operation, installation, and maintenance of the DNA110 SpeedVac. It details the system's functionality in drying low-volume ethanol or isopropanol precipitates of DNA/RNA using centrifugal force and vacuum. Key features include an integral oil-free vacuum pump, chamber heater with adjustable drying rates, and available rotors for various tube sizes. Accessories like a chemical trap kit for solvent vapors are also discussed.
Description:
Instruction manual
Environment:
Savant DNA110 SpeedVac Concentrator, input power:
DNA110
DNA-110
DNA 110
- 120 VAC / 60 Hz, 10A
- 240 VAC / 50 Hz, 5A
Attachment(s):
| File | Last Modified |
|---|---|
| 104-3001-00 - Rev E - Savant DNA110 SpeedVac Concentrator - Instruction Manual.pdf | January 03, 2022 |