Instruction Manual - Digital Vacuum Gauge - DVG50
The Digital Vacuum Gauge DVG50 instruction manual provides comprehensive guidance on installation and operation. It covers receiving and inspecting the gauge, power requirements (15 VDC, 400 mA), and gauge tube installation, emphasizing vertical orientation to prevent particle accumulation. Operational instructions detail reading vacuum levels in Torr or mTorr, and the impact of thermal conductivity on calibration in various environments.
Description:
DVG 50 manual