Savant AES1000 and AES2000 Automatic Environmental SpeedVac with VaporNet - Instruction Manual
The Savant AES1000 and AES2000 Automatic Environmental SpeedVac with VaporNet - Instruction Manual provides comprehensive guidance on using these vacuum concentrators. It covers installation, operation, and specifications, detailing the capacities and power requirements for both models. The AES1000 is designed for moderate capacity with options for different voltages, while the AES2000 supports larger capacities. Both models integrate VaporNet technology for efficient solvent recovery.
Description:
Instruction manual
Environment:
Savant Automatic Environmental SpeedVac with VaporNet, models:
| AES1000 | AES2000 | |
| Capacity: | Moderate | Large |
| Operative Power: | 120 VAC; 60 Hz; 12 amps | 220 VAC; 60 Hz; 8 amps |
| 240 VAC; 50 Hz; 6 amps | 240 VAC; 50 Hz; 8 amps |
AES1000-115
AES1000-220
AES2000-220
AES2000-240
*Depends on ambient temperature, line voltage fluctuations, and load capacity.
Attachment(s):
| File | Last Modified |
|---|---|
| Savant AES1000 and AES2000 Automatic Environmental SpeedVac with VaporNet - Instruction Manual.pdf | January 03, 2022 |