AN-IODWINE-E 0315 - Rev A - AN 016 Standardization of Iodine Titrant for Ripper Titration of Wine
This application note outlines the process for standardizing iodine titrant in Ripper titration for wine analysis. It highlights the importance of determining the true concentration of the iodine titrant using a standard sodium thiosulfate solution and a Thermo Scientific Orion electrode. The workflow includes preparing titrant and standard solutions, performing the titration, and calculating the normality of the iodine titrant. Optional quality control samples for sulfur dioxide (SO2) are also
Description:
Application note for performing the standardization of iodine titrant for Ripper titration of wine.
Attachment(s):
| File | Last Modified |
|---|---|
| AN-IODWINE-E 0315 - Rev A - AN 016 Standardization of Iodine Titrant for Ripper Titration of Wine.pdf | November 10, 2023 |