AN-PLTORPWINE-E 0816 - Rev B - AN 018 Sulfur Dioxide in Wine by Manual Ripper Titration with a Platinum ORP Electrode
This article explains how to analyze sulfur dioxide (SO2) in wine using manual Ripper titration with a Thermo Scientific Orion platinum ORP electrode. It details the necessary equipment, outlines the titration process, and provides calculations for SO2 concentration. Quality control recommendations and electrode maintenance tips are included. The method aims to improve accuracy and eliminate color interference in red wine analysis
Description:
Application note for performing the determination of sulfur dioxide in wine by manual Ripper titration with a platinum ORP electrode.
Attachment(s):
| File | Last Modified |
|---|---|
| AN-PLTORPWINE-E 0816 - Rev B - AN 018 Sulfur Dioxide in Wine by Manual Ripper Titration with a Platinum ORP Electrode.pdf | November 10, 2023 |