AN-PUREWATER-E 0617 - Rev C - AN 002 Conductivity of Pure Water by USP 645
This article details measuring pure water conductivity using USP<645> standards. It lists required equipment like Thermo Scientific™ Orion™ meters and electrodes and outlines the three testing stages. It provides instructions for sample preparation, meter setup, electrode calibration, and analysis. Emphasis is placed on corrective actions, quality control, and proper equipment cleaning/storage to ensure accurate water purity measurement, essential for pharmaceutical and food/beverage industries
Description:
Application note for testing of pure water conductivity by USP 645 guidelines.
Environment:
- 013016MD (2-Cell Conductivity Probe for Ultra Pure Water)
Attachment(s):
| File | Last Modified |
|---|---|
| AN-PUREWATER-E 0617 - Rev C - AN 002 Conductivity of Pure Water by USP 645.pdf | December 13, 2022 |