AN-SALTCAN-E 0215 - RevA - AN 014 Measuring Salt Content in Canned Vegetables Using Manual Titration
This article outlines measuring salt content in canned vegetables using manual titration with Thermo Scientific Orion Chloride Ion Selective Electrode (ISE) or Silver Billet Electrode. It details the needed equipment, reagent preparation, meter setup, and titration process. The method includes electrode setup, sample preparation, titrant standardization, and calculations. Results show manual titration accuracy comparable to autotitration and ion chromatography.
Description:
Application note for measuring salt content in canned vegetables using manual titration.
Environment:
9617BNWP Orion Chloride ISE Electrode
9780SC Orion Silver Billet Electrode
Attachment(s):
| File | Last Modified |
|---|---|
| AN-SALTCAN-E 0215 - RevA - AN 014 Measuring Salt Content in Canned Vegetables Using Manual Titration.pdf | May 31, 2024 |