S-STARA112-E 1213 - RevB - Orion Star A112 Cond Meter Product Specifications
The Thermo Scientific Orion Star A112 Conductivity Benchtop Meter offers a blend of simplicity and precision, featuring a large LCD for clear conductivity/TDS readings and temperature, along with battery life and reading stability indicators. With AUTO-READ™, automatic/manual calibration, and auto-ranging measurements, it ensures high accuracy. The meter supports various power sources and includes an electrode arm and holder. It’s IP54-rated for splash resistance and can log up to 50 data points
Description:
Product specification sheet for STARA112.
Environment:
- STARA112
Attachment(s):
| File | Last Modified |
|---|---|
| S-STARA112-E 1213 - RevB - Orion Star A112 Cond Meter Product Specifications.pdf | November 10, 2022 |