S-STARA113-E 1213 - RevB - Orion Star A113 DO Meter Product Specifications
The Thermo Scientific Orion Star A113 Dissolved Oxygen Benchtop Meter provides an affordable way to measure dissolved oxygen. Features include a large screen for DO readings and temperature, automatic temperature compensation, manual salinity and barometric pressure corrections, and memory for 50 data points. It supports two calibration methods, accepts Orion polarographic DO probes, and can be powered by an adapter or AA batteries. The IP54-rated housing is splash-resistant and wall-mountable.
Description:
Product specification sheet for STARA113.
Environment:
- STARA113
Attachment(s):
| File | Last Modified |
|---|---|
| S-STARA113-E 1213 - RevB - Orion Star A113 DO Meter Product Specifications.pdf | November 10, 2022 |