Thermo Fisher Scientific
AccumetComm Computer Software
To communicate with certain Accumet meters, you need to use compatible software designed for your computer's system (either 32-bit or 64-bit). Ensure you are using the correct version. Compatible models include Accumet AB150, AB200, AB250, XL150, XL200, XL250, XL500, and XL600. However, the following models are not compatible: AB15/AB15Plus, AB315, AB330, XL15, XL20, XL25, XL30, XL40, XL50, XL60, and any older Accumet models.
Description: 
Software to be used on a computer to communicate with certain Accumet meters.
Be sure to use the correct version for your system (either 32-bit or 64-bit).
Compatible meters include Accumet models AB150, AB200, AB250, XL150, XL200, XL250, XL500, and XL600.
The following Accumet models are NOT compatible: AB15/AB15Plus, AB315, AB330, XL15, XL20, XL25, XL30, XL40, XL50, XL60, and any older Accumet models.
  • Was this article helpful?