3177876 - Rev M - Coliform Incubator Bath - Operating and Maintenance Manual
The Coliform Incubator Bath Operating and Maintenance Manual (document 3177876 - Rev M) provides comprehensive guidance for the installation, operation, and maintenance of the Coliform Incubator Bath. This manual ensures users can effectively install, operate, and maintain their Coliform Incubator Bath.
Description:
Operating and maintenance manual
Environment:
Coliform Incubator Bath model 2862/2863 (253):
| Models covered by this manual: | ||||||
| Catalog No. | Volts | Hertz | Watts | Amps | Mains Fuse | Power Board Fuse |
| 3166678 | 120 | 50/60 | 1550 | 12.9 | N/A | N/A |
| 3166679 | 240 | 50/60 | 1550 | 6.5 | FST-5x20-10A | FST-6.3x32-6.3mA |
Attachment(s):
| File | Last Modified |
|---|---|
| 3177876 - Rev M - Coliform Incubator Bath - Operating and Maintenance Manual.pdf | December 01, 2021 |