3177877 - Rev I - Microprocessor-Controlled General Purpose Water Baths - Installation and Operation Manual
The Microprocessor-Controlled General Purpose Water Baths Installation and Operation Manual (document 3177877 - Rev I) provides detailed instructions for the installation, operation, and maintenance of these water baths. This manual ensures users can effectively install, operate, and maintain their microprocessor-controlled general-purpose water baths.
Description:
Installation and operation manual
Environment:
Microprocessor-Controlled General Purpose Water Baths, models:
- 280 (2825/2826)
- 281 (2829/2830)
- 282 (2833/2834)
- 283 (2837/2838)
- 284 (2841/2842)
- 285 (2845/2846)
- 286 (2849/2850)
- 288 (2853/2854)
| Table 3.1 Listing of Models Included in this Manual | |||||||
| Model Number | Catalog Number | Electrical Characteristics | Capacity | 240V Units Mains Fuse | |||
| Volts | Watts | Amps | Liters | Gallons | |||
| 2825 | 3166686 | 120 | 225 | 1.9 | 1.5 | 0.4 | FST-5x20-1 .OA |
| 2826 | 3166687 | 240 | 225 | 1 | 1.5 | 0.4 | FST-5x20-1 .OA |
| 2829 | 3166688 | 120 | 225 | 1.9 | 2.5 | 0.7 | FST-5x20-1 .OA |
| 2830 | 3166689 | 240 | 225 | 1 | 2.5 | 0.7 | FST-5x20-1 .OA |
| 2833 | 3166690 | 120 | 300 | 2.5 | 5.5 | 1.5 | FST-5x20-2.0A |
| 2834 | 3166691 | 240 | 300 | 1.3 | 5.5 | 1.5 | FST-5x20-2.0A |
| 2837 | 3166692 | 120 | 400 | 3.3 | 12 | 3.2 | FST-5x20-2.0A |
| 2838 | 3166693 | 240 | 400 | 1.7 | 12 | 3.2 | FST-5x20-2.0A |
| 2841 | 3166694 | 120 | 600 | 5 | 19.5 | 5.2 | FST-5x20-3.15A |
| 2842 | 3166695 | 240 | 600 | 2.6 | 19.5 | 5.2 | FST-5x20-3.15A |
| 2845 | 3166696 | 120 | 600 | 5 | 18 | 4.9 | FST-5x20-3.15A |
| 2846 | 3166697 | 240 | 600 | 2.6 | 18 | 4.9 | FST-5x20-3.15A |
| 2849 | 3166698 | 120 | 1200 | 10 | 43 | 11.4 | FST-5x20-6.3A |
| 2850 | 3166699 | 240 | 1200 | 5.2 | 43 | 11.4 | FST-5x20-6.3A |
| 2853 | 3166700 | 120 | 800 | 6.7 | 12.0* | 3.2* | FST-5x20-3.15A |
| 2854 | 3166701 | 240 | 800 | 3.5 | 12.0* | 3.2* | FST-5x20-3.15A |
| * Each chamber | |||||||
| For all 240V models: Power Supply Board Fuse - FST-5x20-6.3mA | |||||||
Attachment(s):
| File | Last Modified |
|---|---|
| 3177877 - Rev I - Microprocessor-Controlled General Purpose Water Baths - Installation and Operation Manual.pdf | December 01, 2021 |