3177889 - Rev G - Thermo Scientific Precision General Purpose Water Baths - Operating and Maintenance Manual
The Thermo Scientific Precision General Purpose Water Baths Operating and Maintenance Manual (document 3177889 - Rev G) provides detailed guidance on the installation, operation, and maintenance of these water baths. This manual ensures users can effectively install, operate, and maintain their Thermo Scientific Precision General Purpose Water Baths.
Description:
Operating and maintenance manual
Environment:
Thermo Scientific Precision General Purpose Water Baths Model 180 Series, models covered:
- 180 (2823/2824)
- 181 (2827/2828)
- 182 (2831/2832)
- 183 (2835/2836)
- 184 (2839/2840)
- 185 (2843/2844)
- 186 (2847/2848)
- 188 (2851/2852)
| Catalog Number | Model Number | Electrical Characteristics | Capacity | 230V Units Power Supply Mains Fuse | |||
| Volts | Watts | Amps | Liters | Gallons | |||
| 3166711 | 2823 | 120 | 225 | 1.9 | 1.5 | 0.4 | FST-5x20-1.0A |
| 3166712 | 2824 | 230 | 225 | 1.0 | 1.5 | 0.4 | FST-5x20-1.0A |
| 3166706 | 2827 | 120 | 225 | 1.9 | 2.5 | 0.7 | FST-5x20-1.0A |
| 3166821 | 2828 | 230 | 225 | 1.0 | 2.5 | 0.7 | FST-5x20-1.0A |
| 3166714 | 2831 | 120 | 300 | 2.5 | 5.5 | 1.5 | FST-5x20-2.0A |
| 3166709 | 2832 | 230 | 300 | 1.3 | 5.5 | 1.5 | FST-5x20-2.0A |
| 3166702 | 2835 | 120 | 400 | 3.3 | 12 | 3.2 | FST-5x20-2.0A |
| 3166704 | 2836 | 230 | 400 | 1.7 | 12 | 3.2 | FST-5x20-2.0A |
| 3166716 | 2839 | 120 | 600 | 5.0 | 19.5 | 5.2 | FST-5x20-3.15A |
| 3166710 | 2840 | 230 | 600 | 2.6 | 19.5 | 5.2 | FST-5x20-3.15A |
| 3166707 | 2843 | 120 | 600 | 5.0 | 18.0 | 4.9 | FST-5x20-3.15A |
| 3166708 | 2844 | 230 | 600 | 2.6 | 18.0 | 4.9 | FST-5x20-3.15A |
| 3166713 | 2847 | 120 | 1200 | 10.0 | 43.0 | 11.4 | FST-5x20-6.3A |
| 3166715 | 2848 | 230 | 1200 | 5.2 | 43.0 | 11.4 | FST-5x20-6.3A |
| 3166703 | 2851 | 120 | 800 | 6.7 | 12 | 3.2 | FST-5x20-4.0A |
| 3166705 | 2852 Dual | 230 | 800 | 3.5 | both pans | both pans | FST-5x20-4.0A |
The 240V units described in this manual were designed specifically for the European market and are supplied with a European style power cord. for domestic use, a U.S. style power cord (P/N 3176836) must be ordered separately.
Attachment(s):
| File | Last Modified |
|---|---|
| 3177889 - Rev G - Thermo Scientific Precision General Purpose Water Baths - Operating and Maintenance Manual.pdf | December 01, 2021 |