3177890 - Rev N - Thermo Scientific Precision Circulating Water Baths - Operating and Maintenance Manual
The Thermo Scientific Precision Circulating Water Baths Operating and Maintenance Manual (document 3177890 - Rev N) provides detailed instructions for the installation, operation, and maintenance of these water baths. This manual ensures users can effectively install, operate, and maintain their Thermo Scientific Precision Circulating Water Baths.
Description:
Operating and maintenance manual
Environment:
Thermo Scientific Precision Circulating Water Baths models 260, 265 and 270:
| Models Covered: | |||||
| Catalog No. | Models | Voltage | Hz | Watts | Amps |
| 3166680 | 2864 (260) | 120 | 50/60 | 1050 | 8.8 |
| 3166682 | 2865 (260) | 230 | 50/60 | 1050 | 4.4 |
| 3166684 | 2866 (265) | 120 | 50/60 | 1550 | 12.9 |
| 3166685 | 2867 (265) | 230 | 50/60 | 1550 | 6.5 |
| 3166681 | 2868 (270) | 120 | 50/60 | 1550 | 12.9 |
| 3166683 | 2869 (270) | 230 | 50/60 | 1550 | 6.5 |
Note: The 230V units described in this manual were designed specifically for the European market and are supplied with a European style power cord. For domestic use, a U.S. style power cord (P/N 3176836) must be ordered separately.
Attachment(s):
| File | Last Modified |
|---|---|
| 3177890 - Rev N - Thermo Scientific Precision Circulating Water Baths - Operating and Maintenance Manual.pdf | December 01, 2021 |