Thermo Fisher Scientific
3177892 - Rev K - Reciprocal Shaking Water Baths - Operating and Maintenance Manual
The Reciprocal Shaking Water Baths Operating and Maintenance Manual (document 3177892 - Rev K) provides comprehensive instructions for the installation, operation, and maintenance of these water baths. This manual ensures users can effectively install, operate, and maintain their Reciprocal Shaking Water Baths.
Description: 
Operating and maintenance manual
Environment:
Reciprocal Shaking Water Baths Model 2870/2871 (25) and 2872/2873 (50):
Table 1. Listing of Models included in this Manual
Model # Volts Hertz Watts Amps Mains Fuse Power Supply Board Fuse
2870 120 50/60 1050 8.8 N/A N/A
2871 240 50/60 1050 4.4 FST-5x20-6.3A FST-6.3 x 32-63mA
2872 120 50/60 1550 12.9 N/A N/A
2873 240 50/60 1550 6.5 FST-5x20-10A FST-6.3 x 32-63mA