ANWPS2PBACT 1014 - Thermo Scientific Barnstead Smart2Pure Water Purification System - Determination of Bacteria Retention
The article assesses the Thermo Scientific Barnstead Smart2Pure water purification system's ability to retain bacteria, using the European Pharmacopoeia's membrane filtration method. It underscores the need for bacteria-free water in labs to avoid experimental variability. Tests showed the Smart2Pure system effectively reduced tap water bacterial load to less than 0.01 CFU/ml. The system uses a 0.2 µm filter and multiple purification steps, ensuring high-quality lab water.
Description:
Determination of bacteria retention
Environment:
Thermo Scientific Barnstead Smart2Pure Water Purification System
Attachment(s):
| File | Last Modified |
|---|---|
| ANWPS2PBACT 1014 - Thermo Scientific Barnstead Smart2Pure Water Purification System - Determination of Bacteria Retention.pdf | January 07, 2022 |