Customer Communication - Shelf Life Extended for Select Filter Cartridges - May 2018
Thermo Fisher Scientific™ has extended the shelf life of select filter cartridges to 24 months from the manufacturing date, based on recent documentation updates and analysis. This extension applies to both existing unopened cartridges in the field and inventory. Shelf life refers to unopened cartridges, while service life pertains to installed cartridges, dependent on various factors such as feed water quality and usage volume.
Description:
This notification is to update customers on the shelf life extension for select filter cartridges. Noted filter cartridges will have their shelf life extended to 24 months from the manufacturing date, instead of the existing 12 months.
Environment:
affected filter part numbers
09.1006
09.1018
09.1019
09.1020
09.1102
09.2003
09.2005
09.2006
09.2012
09.2201
09.4011
Attachment(s):
| File | Last Modified |
|---|---|
| Thermo Fisher Scientific IX Cartridges Extended SL - Rev3.pdf | February 14, 2023 |