Purity of ultrapure water in a beaker does not match the purity on the water system display
This article addresses the misconception that a discrepancy between the purity of ultrapure water in a beaker and the water system display indicates a problem with the water system. The purity can change rapidly after being dispensed due to CO2 absorption, forming carbonic acid and lowering purity. To verify water purity accurately, it is best to use an inline conductivity meter with a flow cell. Consistent maintenance of the water system is recommended to avoid inaccurate results and downtime
Issue
When I check the purity of ultrapure water in a beaker, it does not match the purity on the water system display. Is there a problem with my water system? How can I verify the purity on my display is correct?
Environment
Ultrapure water purification
Resolution
Refer to the attached document
Attachment(s)
| File | Last Modified |
|---|---|
| Purity of ultrapure water in a beaker does not match the purity on the water system display.pdf | January 14, 2022 |