Service Instructions - Aquanex Control Panel and Top Cover Removal Video
This article has a video that shows the step required for an authorized service technician to gain access to the control board. This is done by removing the external covers, which is shown in the video instructions. You remove the screws, then gently pry the plastic panel off being careful not the damage the clips of each panel. Tank vent filter must be removed to gain access to the circuit board.
Topic
Short video (no audio) instructions for removing the top control panel and top cover of the Aquanex™ pure water system. Has helpful info for accessing main board.
Environment
7101010 Aquanex 10L System with Dispenser
7101011 Aquanex 10L System with Dispenser and Pretreatment
7201030 Aquanex 30L System with Dispenser and 30L tank
7201031 Aquanex 30L System with Dispenser, 30L tank and Pretreatment