Barnstead - Concentrate Flow Adjustments Instruction Sheet
This article explains adjusting the concentrate flow in reverse osmosis systems, defining terms like concentrate, permeate, CCW, and CW, and listing necessary tools. It stresses the importance of adjustments after installation and every three months for optimal water production and membrane life. Using the "36 Second" Rule for measurement, it lists models needing regular adjustments: Barnstead Smart2Pure, Pacific TII, LabTower RO, Pacific RO, LabTower EDI, and LabTower TII
Description:
Concentrate Flow Adjustment instructions. Recommended procedure following replacement of RO membrane or installation of new RO system.
Environment:
Models:
- Barnstead Pacific TII
- Barnstead Pacific RO
- Barnstead Smart2Pure
- Barnstead LabTower EDI
- Barnstead LabTower RO
- Barnstead LabTower TII
Attachment(s):
| File | Last Modified |
|---|---|
| Concentrate Flow Adjustments rev 4.pdf | March 01, 2023 |