Barnstead Nanopure - How to change the Pressure Reducing Valve
This article provides a step-by-step guide for replacing the pressure-reducing valve in a unit. It details the process from depressurizing and unplugging the unit, removing the back cover, locating and marking the tubes, disconnecting and removing the old valve, installing the new valve, reconnecting the tubes, and finally reassembling and checking the unit for leaks.
Description:
How to change the Pressure Reducing Valve, PM1190X1
Environment:
Barnstead Nanopure
Attachment(s):
| File | Last Modified |
|---|---|
| Barnstead Nanopure - How to change the Pressure Reducing Valve.pdf | January 14, 2022 |