Genpure - Low Purity Troubleshooting Guide
This Troubleshooting Guide is for Genpure Water Systems experiencing Low Purity. If the resistivity level on a Genpure water purification system is too far below 18.0 Mohm-cm, follow the steps provided: check installation, estimate water usage, ensure Nonstop mode, verify cartridge age, enable temperature compensation, check water temperature, inspect rinse line, verify feed water quality, measure flow rate, observe purity reading during dispensing, and check calibration constants.
Description:
Troubleshooting guide for purity reading not reaching desired levels
Environment:
GenPure
Attachment(s):
| File | Last Modified |
|---|---|
| Genpure - Low Purity Troubleshooting Guide.pdf | January 05, 2022 |