Thermo Fisher Scientific
Replacement Threaded Stopper for Disinfection Cartridges
This article provides part numbers for the replacement threaded plug for reusable disinfection cartridges used in Genpure and Micropure water purification systems. Disinfection cartridge 09.2201 is for Genpure systems, and 09.1102 is for Micropure systems. Part# 14.0091 is for cartridges made before 2017, while Plug part# 880052 and O-ring part# 101029 are for units from 2017 to the present.
Description: 
The attached document documents the two different part numbers for the replacement threaded plug on the reusable disinfection cartridges. The disinfection cartridge is an accessory required to clean the water purification system.  If a replacement cap is required, the part number depends on the age. The correct part number can be determined by the appearance.
Environment:
disinfection cartridge 09.2201 - for Genpure water purification systems
disinfection cartridge 09.1102 – for Micropure water purification systems
  • Was this article helpful?