Thermo Fisher Scientific
Heracell Incubator - How to set up the Telemetry Cable Set Instruction Sheet
This instruction sheet provides detailed steps for setting up the telemetry cable for Heracell Incubators. It specifies changing the BAUD rate to 9600 on both the incubator and DLC, navigating interface settings, and adding sensor details such as type, USB port, baud rate, and voltage. Users must also enter account email and device serial number, then deploy changes to save settings.
Description: 
How to set up the Telemetry Cable Set for Heracell Incubator to use with devicelink connect. Linking to instrument health via DLC requires Baud rate settings change on incubator. 
Environment:
DLC, DeviceLink connect, Instrument Health, data push smart cable Heracell incubator, 328928H03 (DLCPTDATA103) 150i /240i, Baud Rate
  • Was this article helpful?