Biosafety Enclosure, Fan Replacement - DT00313 - Rev A
Issue
- Fan RPM is less than 2800 when set to 100%
- Main Vel is less than (Main Vel Calibrate) + 0.020
Environment
All Bigfoot Spectral Cell Sorter Models
Resolution
See attachment for complete resolution.
Attachment(s)
File | Last Modified |
---|---|
DT00313_Fan Replacement, Biosafety Enclosure.pdf | December 08, 2023 |