Biosafety Systems Troubleshooting - DT00314 - Rev A
Issue
-
Aerosol Management System fan is inoperable, doesn't respond to user input, or doesn't maintain 6000 rpm
-
Aerosol Management System HEPA filter fails 3rd party efficiency/integrity testing
-
Biosafety Enclosure fan is inoperable, doesn't respond to user input, doesn't maintain 2800 rpm, or is annoyingly loud
-
Biosafety Enclosure HEPA filter fails 3rd party efficiency/integrity testing
-
Both fans/ all pressure sensors unresponsive
-
Excessively difficult or impossible to balance inflow/downflow
-
Greater than 16 fpm range for downflow from left to right side.
-
Inflow/downflow balance point found at not recommended damper position
-
Liquid leaking out of catch pan instead of draining into instrument drip pan
-
Low remaining HEPA filter remaining life/ Time to replace the HEPA filter warning
-
Non responsive pressure sensors
-
Non sensical pressure sensor values
-
Periodic containment/ low inflow air velocity alarms
-
Select components unresponsive when electrical cables, continuity, and the device is confirmed to be good
Environment
All Bigfoot Spectral Cell Sorter Models
Resolution
See attachment for complete resolution
Attachment(s)
File | Last Modified |
---|---|
DT00314_Biosafety Systems Troubleshooting.pdf | December 08, 2023 |