Boost Pressure and Time - DT00229 - Rev A - Field Calibration
Issue
•A new nozzle tip definition is set up or a new sample line was installed.
•Setting the flow rate to 30%, Bigfoot QC beads take longer than 30 seconds to achieve 300 eps +/- 50
•Post sort analysis shows low recovery.
•Abnormal bead consumption in QC processes (system goes through over a bottle each week).
•Drop delay or bead alignment fails event rate validation.
Environment
All Bigfoot Spectral Cell Sorters
Resolution
See attachment for complete resolution
Attachment(s)
File | Last Modified |
---|---|
DT00229 - Field Calibration - Boost Pressure and Time.pdf | May 05, 2023 |