HEPA Filter Replacement, Biosafety Enclosure - DT00307 - Rev A
Issue
- The main biosafety enclosure fan must operate at 85% or greater to maintain adequate inflow air velocity
-
The HEPA filter is over 10 years old.
-
A HEPA filter integrity test was performed on the HEPA filter, and it failed to reach its required efficiency. This is a part of testing DT00149.
Environment
All Bigfoot Spectral Cell Sorter Models
Resolution
See attachment for complete resolution
Attachment(s)
File | Last Modified |
---|---|
DT00307_HEPA Filter Replacement, Biosafety Enclosure.pdf | August 21, 2023 |