High Pressure Peristaltic Pump Replacement - DT00216 - Rev B - Field Repair
Issue
Fluidic Errors:
•Sheath Air Maintain Failure
•Sheath Level Maintain Failure
•Startup Set Sheath Level
•PreBottle Level Maintain Failure
•PreBottle Air Maintain Failure
•Sample Air Maintain Failure
•DI Pressure Maintain Failure
•Grinding noises from left side of instrument
Failed Tests:
- Pump Test
Environment
All Bigfoot Spectral Cell Sorter Models
Resolution
See attachment for full instructions.
Attachment(s)
File | Last Modified |
---|---|
DT00216 -Field Repair - Replace High Pressure Peristaltic Pump.pdf | May 18, 2023 |