Sample Pressure Offset - DT00228 - Rev A - Field Calibration
Issue
- Sample event rate seems very low, even at high flow rates (compared to other instruments).
- A new nozzle tip definition is set up or a new sample line was installed.
- Drop delay or bead alignment fails event rate validation.
Environment
All Bigfoot Spectral Cell Sorter Models
Resolution
See attachment for complete resolution
Attachment(s)
File | Last Modified |
---|---|
DT00228 - Field Calibration - Sample Pressure Offset.pdf | May 04, 2023 |