Scatter Alignment, Fine Tuning - DT00298 - Rev A - Field Repair
Issue
•Poor Separation of Live/Dead in FSC v SSC plots
•Populations move with Drops turning ON
•BLR (Baseline Restore) warnings on the Scatter channels
•Customer is concerned on the resolution of FSC, SSC, or SPD.
•Drop drive noise appears in the Trigger Plot high enough to interfere with sample detection.
Environment
All Bigfoot Spectral Cell Sorter Models
Certain steps will only pertain to models with a SPD option
Resolution
See attachment for complete resolution
Attachment(s)
File | Last Modified |
---|---|
DT00298 - Field Repair - Fine Tuning Scatters.pdf | August 17, 2023 |