Sort/Nozzle Door Replacement - DT00263 - Rev A - Field Repair
Issue
•Broken door open/closed sensors
•Broken sort door motor
•Bad linear rail
•Broken push to close latch on Nozzle Door
•Broken pulley belt
•Damaged Sort or Nozzle Door
•Obstructed door movement
•Door fails to open/close smoothly
•Door sensors fail to report door open/close properly
•Door fails to open/close completely
Environment
All Bigfoot Spectral Cell Sorter Models
Resolution
See attachment for complete resolution
Attachment(s)
File | Last Modified |
---|---|
DT00263 - Replace Sort-Nozzle Door.pdf | May 19, 2023 |