Thermo Fisher Scientific
633nm HeNe laser front aperture check
Issue
When installing the 633 nm HeNe laser, the output laser power after alignment and calibration should be 8.0 mW. An aperture at the front of the laser output should be checked if the output power can not reach 8.0 mW during installation.
Environment 
  • Raman DXR2
  • Raman DXR3
  • SmartRaman DXR2
  • SmartRaman DXR3
  • Raman DXR2xi
  • Raman DXR3xi
Resolution 
Check and fully open the laser front aperture.
1. Turn off the instrument. Unplug the power cable of the 633 nm laser from the instrument.
2. Remove the long laser tube cover. 
3. At the front of the laser output, there is a tab labeled "BEAM (Open, Closed)". Move the tab to fully OPEN to open the aperture in front of the laser. 
clipboard_e52adffecb0efbed6a97e2726ce77ac4f.png
clipboard_eb8793ff7d68bef0108d4d56547d2cd1c.png
4. When attaching the laser tube cover back, make sure that the foam under the laser tube cover does not rub on the aperture tab to accidently move it to Close or partially close.
clipboard_ea5a7f1415340101fbf84f47674bb369f.png
5. Plug in the power cable. Turn on the instrument and realign/re-calibrate.
  • Was this article helpful?