Alignment and Calibration Frequency
Issue
How often should you align and calibrate a DXR2 or DXR2xi?
Environment
- Raman DXR2
- SmartRaman DXR2
- Raman DXR2xi
Resolution
- You should align a DXR2 microscope weekly.
- You should align a DXR2 Smart Raman biweekly
- You should calibrate a DXR2 microscope daily in a regulated environment and weekly in a non-regulated environment. You should calibrate a DXR2 Smart Raman biweekly.
- You should align and calibrate a DXR2xi daily in a regulated environment and weekly in a non-regulated environment.
The table below summarizes the recommended alignment and calibration frequencies.
| DXR2 | SmartRaman DXR2 | DXR2xi | |
| Alignment Frequency | Weekly | Biweekly |
Daily in a regulated environment Weekly in a non-regulated environment |
| Calibration Frequency |
Daily in a regulated environment Weekly in a non-regulated environment |
Biweekly |
Daily in a regulated environment Weekly in a non-regulated environment |