269-344500 - Rev A - DXR Microscope Addendum - Alignment Tool Orientation
Description:
When installing a DXR or DXRxi microscope, position this updated alignment and calibration tool (part numbers 840-3276 and 840-3277) so that the connecting cable extends toward to microscope, as shown.
If the tool is installed in any other orientation, installation tests will fail.
Environment:
DXR microscopes
DXRxi microscopes
Attachment(s):
| File | Last Modified |
|---|---|
| 344500 A DXR MICROSCOPE ADDENDUM.pdf | March 23, 2023 |