Audit Log Service Database Configuration
Issue
Error message:
Unable to send the new configuration to the service
Environment
- Observed during configuration of the Audit Manager Database in the Security Suite installation procedure
- FT-IR
Resolution
When configuring the Audit Manager Database in the Security Suite installation procedure the step to choose a database engine will ask the end user to select a database engine. If using the SQLite database engine (available for Single Computer configurations only), the Security Suite creates the Audit Manager database automatically and sets appropriate access rights for the Audit Log Service account you specify. No other database is required as the Database Engine is a free data base of Windows. The error occurs when you choose the SQLite database with a distributed install.