Thermo Fisher Scientific
Part Number for Smart iTR Pressure Tip for 3-Bounce Crystal Plates
Issue
The 3-bounce crystal plates for the Smart iTR accessory use a different pressure tip that has a larger Teflon end.
Environment
Smart iTR accessory
Resolution
The part number for the replacement tip is PKE025-3094