Thermo Fisher Scientific
What is the power of the IR energy at the sample compartment of the Nicolet iS50?
Issue
What is the power of the IR energy at the sample compartment of the Nicolet iS50.
Environment
Nicolet iS50
Resolution
The approximate power of the IR energy at the focal point in the sample compartment is 45 milliwatts.
  • Was this article helpful?