Diffuse Reflectance Accessory Installation and User's Guide
Description:
Diffuse reflectance accessory installation and user's Guide
The Pike Technologies Diffuse Reflection accessory is designed for use in FTIR spectrometers. The design utilizes four beam steering flat mirrors and a high collection angle ellipsoidal reflector to collect the maximum amount of energy from the sample. The PIKE Technologies Diffuse Reflection accessory has been created for high energy throughput and ease of use.
Environment:
FT-IR
Attachment(s):
File | Last Modified |
---|---|
Diffuse Reflectance Accessory Installation and User's Guide.pdf | August 04, 2022 |