Thermo Fisher Scientific
FlexIR - Product Data Sheet
Description: 
The PIKE FlexIR™ near-IR (NIR) Fiber Optic Accessory is an excellent tool for remote and speedy analysis of a wide variety of materials. Powders, plastics, coatings, and liquid samples are readily measured – typically within 30 seconds. The FlexIR accessory is ideal for performing incoming QC of materials used in pharmaceutical, polymer, and chemical manufacturing.
Environment:
FlexIR
FT-IR