JetStream ATR Accessory – Measurements of Liquids under Varying Conditions Product Data Sheet
Description:
JetStream ATR accessory product information
The PIKE Technologies JetStream is a unique ATR accessory optimized for analysis of liquids in static or flow modes at varying pressure.
Environment:
- JetStream ATR
- FT-IR
Attachment(s):
File | Last Modified |
---|---|
JetStream ATR Accessory – Measurements of Liquids under Varying Conditions Product Data Sheet.pdf | August 04, 2022 |