AN 50777 - Dedicated Analytical Solutions for Tablets and Softgels, Transmission Analysis of Solid Dosage Pharmaceuticals
Description:
The majority of commercially available pharmaceutical formulations are tablets and capsules designed to be taken orally. Fourier transform near-infrared (FT-NIR) analysisis a powerful technique for measuring these materials. The advantages of using both the Softgel and StandardTablet detectors on the Nicolet™Antaris™FT-NIR Analyzerwill be discussed in this application note. Diffusereflectance and transmission spectra of tablets and softgel capsules will also be shown.
Environment:
- FT-NIR
- Nicolet Antaris
Attachment(s):
File | Last Modified |
---|---|
AN 50777 - Dedicated Analytical Solutions for Tablets and Softgels, Transmission Analysis of Solid Dosage Pharmaceuticals.pdf | July 28, 2022 |