Replacing the power supply
Time needed: 5 minutes or less
Tools needed: None
Use only Thermo Fisher certified replacement parts. To order, contact us. |
Avoid shock hazard. Do not attempt to remove the cover of the power supply. Before you replace the power supply, always make sure you have turned off your instrument and disconnected the power supply from the wall outlet or power strip. |
- Turn off the instrument.
- Disconnect the old power supply (simply pull out the power connector).
- Connect the new power supply and turn on the instrument.
If your instrument does not function normally, turn off the power and check the cable connections between the power supply, the instrument, and the wall outlet or power strip. If the connectors are seated properly and the instrument still does not function normally, contact us for assistance.