Replacing the humidity indicator
The humidity indicator must be replaced when it turns white or does not return to blue after the desiccant has been replaced.
Time needed: Less than 1 minute
Tools needed:
- Gloves, finger cots, or laboratory tissue
- Replacement humidity indicator
Use only Thermo Scientific certified replacement parts. To order parts, contact us. |
- Remove the old humidity indicator.
Save the old indicator for reference when you install the new indicator.
Always wear lab gloves or finger cots, or use laboratory tissue, when handling the humidity indicator. Oil or moisture from skin can discolor the indicator. |
When installing the new humidity indicator, make sure the indicator is centered in the holder and that there are no gaps.
-
Peel the backing off of the indicator holder, and then, as shown below, press the round, blue indicator onto the adhesive on the holder.
-
Place the new indicator and holder over the indicator opening, and press down gently to make sure it is sealed.