How do I maintain my spectrometer?
To keep your spectrometer in good working order, check the desiccant indicator frequently and replace the desiccant canisters as soon as they become saturated.
If you want to clean your spectrometer, you can wipe off the exterior of the main cover, but do not allow any moisture to come into contact with the windows. For more information, see “Cleaning your spectrometer” in Spectrometer Help Topics, available through the OMNIC Paradigm software Help menu.