How do I verify performance?
Ensure that your instrument is working well by running PV workflows and checking the system status.
Test | Description |
Nicolet FTIR - Factory Qualification | Runs factory recommended tests and all qualification tests |
Nicolet FTIR - PV Test | Measures basic performance based on factory recommended tests. |
Nicolet FTIR - CP Qualification | Executes the qualification tests as defined in the Chinese Pharmacopoeia. |
Nicolet FTIR - JP Qualification | Executes the qualification tests as defined in the Japanese Pharmacopoeia. |
Nicolet FTIR - PHEUR Qualification | Executes the qualification tests for as defined in the European Pharmacopoeia. |
Nicolet FTIR - USP Qualification | Executes qualification tests as defined in the US Pharmacopoeia. |
To run a qualification or PV workflow
- Right-click the workflow and select Run.
- Follow the on-screen prompts.
System status
The system status icon displays information about your instrument and software services.
Icon | Icon with Security Suite installed | Description |
![]() |
![]() |
The system is connected and all services are running correctly. You are ready to measure and save data. Click the system status icon to view details about the system. |
![]() |
![]() |
There may be a problem with the instrument, such as:
Click the system status icon for more details about the problem. Also visually inspect the instrument and connection. |
![]() |
![]() |
There is a problem with one or more software services. Click the system status icon for details. If the service does not start automatically after a few minutes, restart the computer. |
The instrument's SPV wheel contains NIST-traceable and NG-11 standards.