269-184200 - IR Spectroscopy of Polymers Booklet
Environment
FT-IR Spectroscopy
Resolution
This booklet discusses the various factors related to this analysis from sample preparation to polymer structure and geometric configuration. This booklet begins with a basic review of the types of molecular motions that result in the absorption of Infra Red energy. It next discusses the spectral affects of molecular geometry and stereochemistry, and later discusses the various classes of polymers and includes spectra and spectral interpretation for several polymers.
Attachment(s)
| File | Last Modified |
|---|---|
| 269-184200 - IR Spectroscopy of Polymers Booklet.pdf | December 22, 2021 |