Heated Demountable Liquid Cell - Product Data Sheet
Description:
The PIKE Technologies Heated Demountable Liquid Cell is recommended for qualitative and quantitative analysis of liquid samples where it is necessary to control the temperature of the sample. Temperature range of the accessory is ambient to 200 °C.
Environment:
Heated Demountable Liquid Cell
Attachment(s):
| File | Last Modified |
|---|---|
| Heated Demountable Liquid Cell - Product Data Sheet.pdf | August 04, 2022 |